1-(2,4-Dibromo-6-fluorophenyl)-2,2-difluoroethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
66329-500MG
66329-1G
66329-5G
66329-25G
77332-786EA
233.81
USD
77332-786
77357-048
77368-836
77390-010
1-(2,4-Dibromo-6-fluorophenyl)-2,2-difluoroethanone ≥97%
1-(2,4-Dibromo-6-fluorophenyl)-2,2-difluoroethanone
Precaución: for Research Use Only
Formula:
C₈H₃Br₂F₃O MW: 331.912 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29089497 CAS Number: 2092680-89-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...