1-(2-Bromo-3-ethoxy-6-fluorophenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

66328-1G 66328-5G 66328-25G
77340-930EA 63.86 USD
77340-930 77368-834 77380-962
1-(2-Bromo-3-ethoxy-6-fluorophenyl)-2,2-difluoroethanone ≥97%
1-(2-Bromo-3-ethoxy-6-fluorophenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₁₀H₈BrF₃O₂
MW: 297.069 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762510
CAS Number: 2901106-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR