1-(6-Bromo-2,3-dichlorophenyl)-2,2-difluoroethanone ≥95%
Supplier: AOBChem USA
|
Warning
|
66327-500MG
66327-1G
66327-5G
66327-25G
77332-784EA
266.77
USD
77332-784
77357-046
77368-832
77390-008
1-(6-Bromo-2,3-dichlorophenyl)-2,2-difluoroethanone ≥95%
1-(6-Bromo-2,3-dichlorophenyl)-2,2-difluoroethanone
Precaución: for Research Use Only
Formula:
C₈H₃BrCl₂F₂O MW: 303.916 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34762509 CAS Number: 2807450-58-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...