1-(3-(Benzyloxy)-2-bromo-6-fluorophenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

66321-500MG 66321-1G 66321-5G 66321-25G
77332-780EA 161.71 USD
77332-780 77345-956 77368-826 77390-006
1-(3-(Benzyloxy)-2-bromo-6-fluorophenyl)-2,2-difluoroethanone ≥97%
1-(3-(Benzyloxy)-2-bromo-6-fluorophenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₁₅H₁₀BrF₃O₂
MW: 359.138 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762505
CAS Number: 2807456-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR