2-Bromo-1-(difluoromethyl)-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

66301-500MG 66301-1G 66301-5G
77337-694EA 436.72 USD
77337-694 77357-038 77380-954
2-Bromo-1-(difluoromethyl)-4-methylbenzene ≥97%
2-Bromo-1-(difluoromethyl)-4-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₇BrF₂
Storage Temperature: Ambient
MDL Number: MFCD18391190
CAS Number: 1261876-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR