4-(Difluoromethyl)-2-methoxy-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

66296-1G 66296-5G 66296-25G
77340-800EA 60.77 USD
77340-800 77368-820 77380-952
4-(Difluoromethyl)-2-methoxy-1-methylbenzene ≥97%
4-(Difluoromethyl)-2-methoxy-1-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀F₂O
Storage Temperature: Ambient
MDL Number: MFCD18395575
CAS Number: 1261500-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR