1,3-Dibromo-5-(difluoromethyl)-2-ethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

66284-1G 66284-5G 66284-25G
77341-718EA 101.97 USD
77341-718 77368-818 77380-950
1,3-Dibromo-5-(difluoromethyl)-2-ethoxybenzene ≥97%
1,3-Dibromo-5-(difluoromethyl)-2-ethoxybenzene

Precaución: for Research Use Only

Formula: C₉H₈Br₂F₂O
Storage Temperature: Ambient
MDL Number: MFCD34762488
CAS Number: 2387083-94-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR