4-Bromo-1-(difluoromethyl)-2-isopropylbenzene ≥95%

Supplier: AOBChem USA

Warning

66281-500MG 66281-1G 66281-5G
77339-944EA 688.04 USD
77339-944 77357-036 77380-948
4-Bromo-1-(difluoromethyl)-2-isopropylbenzene ≥95%
4-Bromo-1-(difluoromethyl)-2-isopropylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrF₂
MW: 249.095 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762486
CAS Number: 2901109-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR