2-Bromo-1-chloro-4-(1,1-difluoroethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

66275-500MG 66275-1G 66275-5G
77339-736EA 621.09 USD
77339-736 77357-034 77380-946
2-Bromo-1-chloro-4-(1,1-difluoroethyl)benzene ≥97%
2-Bromo-1-chloro-4-(1,1-difluoroethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₆BrClF₂
Storage Temperature: Ambient
MDL Number: MFCD32662625
CAS Number: 1784994-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR