2-Bromo-4-(difluoromethyl)-1-ethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

66274-500MG 66274-1G 66274-5G 66274-25G
77332-774EA 264.71 USD
77332-774 77357-032 77368-816 77390-000
2-Bromo-4-(difluoromethyl)-1-ethoxybenzene ≥97%
2-Bromo-4-(difluoromethyl)-1-ethoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉BrF₂O
MW: 251.068 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32176995
CAS Number: 2706133-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR