4-Bromo-1-(difluoromethyl)-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

66269-500MG 66269-1G 66269-5G
77336-120EA 375.95 USD
77336-120 77357-028 77380-942
4-Bromo-1-(difluoromethyl)-2-methylbenzene ≥97%
4-Bromo-1-(difluoromethyl)-2-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₇BrF₂
Storage Temperature: Ambient
MDL Number: MFCD18391194
CAS Number: 1254034-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR