1-Bromo-4-(difluoromethyl)-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

66268-500MG 66268-1G 66268-5G 66268-25G
77332-772EA 244.11 USD
77332-772 77357-026 77368-814 77389-998
1-Bromo-4-(difluoromethyl)-2-methylbenzene ≥95%
1-Bromo-4-(difluoromethyl)-2-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₇BrF₂
MW: 221.042 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18391187
CAS Number: 1261570-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR