3-(4-(Benzyloxy)-2-fluorophenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

66219-500MG 66219-1G 66219-5G
77336-264EA 384.19 USD
77336-264 77357-022 77380-936
3-(4-(Benzyloxy)-2-fluorophenyl)pyridine ≥97%
3-(4-(Benzyloxy)-2-fluorophenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₈H₁₄FNO
Storage Temperature: Ambient
MDL Number: MFCD34762462
CAS Number: 2172101-35-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR