3-(2,3-Dichloro-4-methylphenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

66194-500MG 66194-1G
77340-434EA 1054.72 USD
77340-434 77357-018
3-(2,3-Dichloro-4-methylphenyl)pyridine ≥97%
3-(2,3-Dichloro-4-methylphenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₉Cl₂N
MW: 238.113 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762438
CAS Number: 2918828-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR