2,3-Dichloro-4,4'-dimethyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

66186-500MG 66186-1G 66186-5G
77339-696EA 614.91 USD
77339-696 77357-016 77380-932
2,3-Dichloro-4,4'-dimethyl-1,1'-biphenyl ≥97%
2,3-Dichloro-4,4'-dimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₂Cl₂
Storage Temperature: Ambient
MDL Number: MFCD34762430
CAS Number: 2901083-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR