(2-Fuoro-5-methoxy-4'-(trifluoromethyl)-[1,1'-biphenyl]-3-yl)(methyl)sulfane ≥95%
Supplier: AOBChem USA
|
Warning
|
66183-500MG
66183-1G
66183-5G
66183-25G
77332-770EA
230.72
USD
77332-770
77357-012
77368-810
77389-996
(2-Fuoro-5-methoxy-4'-(trifluoromethyl)-[1,1'-biphenyl]-3-yl)(methyl)sulfane ≥95%
(2-Fuoro-5-methoxy-4'-(trifluoromethyl)-[1,1'-biphenyl]-3-yl)(methyl)sulfane
Precaución: for Research Use Only
Formula:
C₁₅H₁₂F₄OS MW: 316.314 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34762427 CAS Number: 2901066-50-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...