(2,3'-Difluoro-5-methoxy-[1,1'-biphenyl]-3-yl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

66182-500MG 66182-1G 66182-5G 66182-25G
77332-768EA 236.9 USD
77332-768 77357-010 77368-808 77389-994
(2,3'-Difluoro-5-methoxy-[1,1'-biphenyl]-3-yl)(methyl)sulfane ≥97%
(2,3'-Difluoro-5-methoxy-[1,1'-biphenyl]-3-yl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₄H₁₂F₂OS
Storage Temperature: Ambient
MDL Number: MFCD34762426
CAS Number: 2901096-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR