(2-Fluoro-4',5-dimethoxy-[1,1'-biphenyl]-3-yl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

66174-500MG 66174-1G 66174-5G
77332-764EA 324.45 USD
77332-764 77357-006 77380-928
(2-Fluoro-4',5-dimethoxy-[1,1'-biphenyl]-3-yl)(methyl)sulfane ≥97%
(2-Fluoro-4',5-dimethoxy-[1,1'-biphenyl]-3-yl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₅H₁₅FO₂S
MW: 278.342 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762418
CAS Number: 2901106-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR