(2,4'-Difluoro-5-methoxy-[1,1'-biphenyl]-3-yl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

66173-500MG 66173-1G 66173-5G 66173-25G
77332-762EA 249.26 USD
77332-762 77357-004 77368-804 77389-990
(2,4'-Difluoro-5-methoxy-[1,1'-biphenyl]-3-yl)(methyl)sulfane ≥97%
(2,4'-Difluoro-5-methoxy-[1,1'-biphenyl]-3-yl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₄H₁₂F₂OS
MW: 266.306 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762417
CAS Number: 2901096-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR