(2-Fluoro-5-methoxy-4'-methyl-[1,1'-biphenyl]-3-yl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

66172-500MG 66172-1G 66172-25G 66172-5G
77332-760EA 307.97 USD
77332-760 77357-002 77389-988 77380-926
(2-Fluoro-5-methoxy-4'-methyl-[1,1'-biphenyl]-3-yl)(methyl)sulfane ≥97%
(2-Fluoro-5-methoxy-4'-methyl-[1,1'-biphenyl]-3-yl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₅H₁₅FOS
MW: 262.342 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762416
CAS Number: 2901098-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR