3-Bromo-4-fluoro-5-(trifluoromethyl)-1,1'-biphenyl ≥97%
Supplier: AOBChem USA
|
Warning
|
66121-500MG
66121-1G
66121-5G
66121-25G
77332-758EA
238.96
USD
77332-758
77357-000
77368-802
77389-986
3-Bromo-4-fluoro-5-(trifluoromethyl)-1,1'-biphenyl ≥97%
3-Bromo-4-fluoro-5-(trifluoromethyl)-1,1'-biphenyl
Precaución: for Research Use Only
Formula:
C₁₃H₇BrF₄ MW: 319.092 g/mol Storage Temperature: Ambient |
CAS Number:
2918967-65-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...