3-(3-Bromo-4-chloro-5-(trifluoromethyl)phenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

66115-500MG 66115-1G 66115-5G
77338-512EA 493.37 USD
77338-512 77356-998 77380-924
3-(3-Bromo-4-chloro-5-(trifluoromethyl)phenyl)pyridine ≥97%
3-(3-Bromo-4-chloro-5-(trifluoromethyl)phenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₆BrClF₃N
MW: 336.535 g/mol
Storage Temperature: Ambient
CAS Number: 2918965-16-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR