3'-Bromo-4'-chloro-5'-(trifluoromethyl)-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

66110-500MG 66110-1G 66110-5G
77337-946EA 453.2 USD
77337-946 77356-996 77380-922
3'-Bromo-4'-chloro-5'-(trifluoromethyl)-[1,1'-biphenyl]-4-carbaldehyde ≥97%
3'-Bromo-4'-chloro-5'-(trifluoromethyl)-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₇BrClF₃O
Storage Temperature: Ambient
CAS Number: 2918963-50-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR