3-Bromo-4-chloro-5-(trifluoromethyl)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

66107-500MG 66107-1G 66107-5G
77337-944EA 453.2 USD
77337-944 77356-994 77380-920
3-Bromo-4-chloro-5-(trifluoromethyl)-1,1'-biphenyl ≥97%
3-Bromo-4-chloro-5-(trifluoromethyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₇BrClF₃
MW: 335.547 g/mol
Storage Temperature: Ambient
CAS Number: 2918959-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR