(3,4-Difluoro-[1,1'-biphenyl]-2-yl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

66093-500MG 66093-1G 66093-5G 66093-25G
77332-756EA 158.62 USD
77332-756 77345-710 77368-800 77389-984
(3,4-Difluoro-[1,1'-biphenyl]-2-yl)(methyl)sulfane ≥97%
(3,4-Difluoro-[1,1'-biphenyl]-2-yl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₃H₁₀F₂S
Storage Temperature: Ambient
MDL Number: MFCD34762379
CAS Number: 2901084-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR