4-(4-Bromo-2-chloro-5-methylphenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

66087-500MG 66087-1G 66087-5G 66087-25G
77332-754EA 249.26 USD
77332-754 77356-992 77368-798 77389-982
4-(4-Bromo-2-chloro-5-methylphenyl)pyridine ≥97%
4-(4-Bromo-2-chloro-5-methylphenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₉BrClN
MW: 282.564 g/mol
Storage Temperature: Ambient
CAS Number: 2918961-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR