4-Bromo-2-chloro-5-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

66079-500MG 66079-1G 66079-5G 66079-25G
77332-752EA 154.5 USD
77332-752 77345-556 77368-796 77389-980
4-Bromo-2-chloro-5-methyl-1,1'-biphenyl ≥97%
4-Bromo-2-chloro-5-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrCl
MW: 281.575 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762377
CAS Number: 2901084-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR