3,4'-Dibromo-4-methyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

66075-500MG 66075-1G 66075-5G 66075-25G
77332-750EA 141.11 USD
77332-750 77344-874 77368-794 77389-978
3,4'-Dibromo-4-methyl-1,1'-biphenyl ≥95%
3,4'-Dibromo-4-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀Br₂
MW: 326.027 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762374
CAS Number: 2901066-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR