3'-Bromo-2,3,4'-trimethyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

66071-500MG 66071-1G 66071-5G 66071-25G
77332-748EA 120.51 USD
77332-748 77343-794 77368-792 77380-918
3'-Bromo-2,3,4'-trimethyl-1,1'-biphenyl ≥97%
3'-Bromo-2,3,4'-trimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₅Br
MW: 275.184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762373
CAS Number: 2807448-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR