3'-Bromo-2,4'-dimethyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

66070-500MG 66070-1G 66070-5G 66070-25G
77332-746EA 136.99 USD
77332-746 77344-622 77368-790 77389-976
3'-Bromo-2,4'-dimethyl-1,1'-biphenyl ≥97%
3'-Bromo-2,4'-dimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃Br
MW: 261.157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762372
CAS Number: 1166838-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR