3-Bromo-4,4'-dimethyl-1,1'-biphenyl ≥97%
Supplier: AOBChem USA
|
Warning
|
66067-500MG
66067-1G
66067-5G
66067-25G
77332-740EA
136.99
USD
77332-740
77344-620
77368-784
77389-970
3-Bromo-4,4'-dimethyl-1,1'-biphenyl ≥97%
3-Bromo-4,4'-dimethyl-1,1'-biphenyl
Precaución: for Research Use Only
Formula:
C₁₄H₁₃Br Storage Temperature: Ambient |
MDL Number:
MFCD34762369 CAS Number: 132462-56-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...