4-Methyl-4'-(trifluoromethoxy)-[1,1'-biphenyl]-3-amine ≥97%

Supplier: AOBChem USA

Warning

66066-500MG 66066-1G 66066-5G 66066-25G
77332-738EA 184.37 USD
77332-738 77347-300 77368-782 77389-968
4-Methyl-4'-(trifluoromethoxy)-[1,1'-biphenyl]-3-amine ≥97%
4-Methyl-4'-(trifluoromethoxy)-[1,1'-biphenyl]-3-amine

Precaución: for Research Use Only

Formula: C₁₄H₁₂F₃NO
MW: 267.246 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12859689
CAS Number: 1184677-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR