4-Methyl-4'-(trifluoromethyl)-[1,1'-biphenyl]-3-amine ≥95%

Supplier: AOBChem USA

Warning

66065-500MG 66065-1G 66065-5G 66065-25G
77332-736EA 184.37 USD
77332-736 77347-298 77368-780 77389-966
4-Methyl-4'-(trifluoromethyl)-[1,1'-biphenyl]-3-amine ≥95%
4-Methyl-4'-(trifluoromethyl)-[1,1'-biphenyl]-3-amine

Precaución: for Research Use Only

Formula: C₁₄H₁₂F₃N
MW: 251.247 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12859574
CAS Number: 1179245-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR