2',4-Dimethyl-[1,1'-biphenyl]-3-amine ≥97%

Supplier: AOBChem USA

Warning

66058-500MG 66058-1G 66058-5G 66058-25G
77332-730EA 272.95 USD
77332-730 77356-988 77368-774 77389-960
2',4-Dimethyl-[1,1'-biphenyl]-3-amine ≥97%
2',4-Dimethyl-[1,1'-biphenyl]-3-amine

Precaución: for Research Use Only

Formula: C₁₄H₁₅N
Storage Temperature: Ambient
MDL Number: MFCD12779860
CAS Number: 1048389-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR