4'-Methoxy-4-methyl-[1,1'-biphenyl]-3-amine ≥97%

Supplier: AOBChem USA

Warning

66056-500MG 66056-1G 66056-5G 66056-25G
77332-728EA 204.97 USD
77332-728 77356-986 77368-772 77389-958
4'-Methoxy-4-methyl-[1,1'-biphenyl]-3-amine ≥97%
4'-Methoxy-4-methyl-[1,1'-biphenyl]-3-amine

Precaución: for Research Use Only

Formula: C₁₄H₁₅NO
MW: 213.275 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12824077
CAS Number: 761440-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR