4,4'-Dimethyl-[1,1'-biphenyl]-3-amine ≥97%

Supplier: AOBChem USA

Warning

66054-1G 66054-5G 66054-25G
77342-030EA 111.24 USD
77342-030 77368-770 77380-916
4,4'-Dimethyl-[1,1'-biphenyl]-3-amine ≥97%
4,4'-Dimethyl-[1,1'-biphenyl]-3-amine

Precaución: for Research Use Only

Formula: C₁₄H₁₅N
Storage Temperature: Ambient
MDL Number: MFCD12824036
CAS Number: 116668-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR