4-Methyl-3-nitro-4'-(trifluoromethoxy)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

66052-500MG 66052-1G 66052-5G 66052-25G
77332-726EA 202.91 USD
77332-726 77356-984 77368-768 77389-956
4-Methyl-3-nitro-4'-(trifluoromethoxy)-1,1'-biphenyl ≥97%
4-Methyl-3-nitro-4'-(trifluoromethoxy)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₀F₃NO₃
MW: 297.229 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18407991
CAS Number: 1261506-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR