3-(4-Methyl-3-nitrophenyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

66048-500MG 66048-1G 66048-5G 66048-25G
77332-718EA 202.91 USD
77332-718 77356-976 77368-762 77389-948
3-(4-Methyl-3-nitrophenyl)pyridine ≥95%
3-(4-Methyl-3-nitrophenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD15476755
CAS Number: 4373-62-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR