4'-Methyl-3'-nitro-[1,1'-biphenyl]-3-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

66046-500MG 66046-1G 66046-5G 66046-25G
77332-716EA 141.11 USD
77332-716 77344-870 77368-760 77389-946
4'-Methyl-3'-nitro-[1,1'-biphenyl]-3-carbaldehyde ≥97%
4'-Methyl-3'-nitro-[1,1'-biphenyl]-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD34762368
CAS Number: 2807446-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR