2,3,4'-Trimethyl-3'-nitro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

66045-500MG 66045-1G 66045-25G 66045-5G
77332-714EA 305.91 USD
77332-714 77356-974 77389-944 77380-912
2,3,4'-Trimethyl-3'-nitro-1,1'-biphenyl ≥97%
2,3,4'-Trimethyl-3'-nitro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD34762367
CAS Number: 2807470-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR