2,4'-Dimethyl-3'-nitro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

66044-500MG 66044-1G 66044-5G 66044-25G
77332-712EA 147.29 USD
77332-712 77345-244 77368-758 77389-942
2,4'-Dimethyl-3'-nitro-1,1'-biphenyl ≥97%
2,4'-Dimethyl-3'-nitro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃NO₂
MW: 227.259 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762366
CAS Number: 854456-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR