4'-Methoxy-4-methyl-3-nitro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

66042-500MG 66042-1G 66042-5G
77335-514EA 356.38 USD
77335-514 77356-972 77380-910
4'-Methoxy-4-methyl-3-nitro-1,1'-biphenyl ≥97%
4'-Methoxy-4-methyl-3-nitro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃NO₃
MW: 243.258 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20483675
CAS Number: 247189-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR