4'-Fluoro-4-methyl-3-nitro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

66041-500MG 66041-1G 66041-5G
77335-450EA 353.29 USD
77335-450 77356-970 77380-908
4'-Fluoro-4-methyl-3-nitro-1,1'-biphenyl ≥97%
4'-Fluoro-4-methyl-3-nitro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀FNO₂
MW: 231.222 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00800375
CAS Number: 205526-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR