4'-Chloro-4-methyl-[1,1'-biphenyl]-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

66007-500MG 66007-1G 66007-5G
77332-708EA 347.11 USD
77332-708 77356-966 77380-904
4'-Chloro-4-methyl-[1,1'-biphenyl]-3-carboxylic acid ≥97%
4'-Chloro-4-methyl-[1,1'-biphenyl]-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₁ClO₂
MW: 246.689 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21508397
CAS Number: 1492861-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR