2-Bromo-4',6-dimethoxy-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

66003-500MG 66003-1G 66003-5G 66003-25G
77332-706EA 232.78 USD
77332-706 77356-962 77368-754 77389-938
2-Bromo-4',6-dimethoxy-1,1'-biphenyl ≥97%
2-Bromo-4',6-dimethoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃BrO₂
MW: 293.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762359
CAS Number: 2807440-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR