1-(2,5-Dichlorophenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

65990-500MG 65990-1G 65990-5G 65990-25G
77332-702EA 258.53 USD
77332-702 77356-958 77368-748 77389-934
1-(2,5-Dichlorophenyl)-2,2-difluoroethanone ≥97%
1-(2,5-Dichlorophenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₈H₄Cl₂F₂O
MW: 225.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26045050
CAS Number: 1564615-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR