1-(3,6-Dichloro-2-fluorophenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

65987-500MG 65987-1G 65987-5G 65987-25G
77332-700EA 258.53 USD
77332-700 77356-956 77368-746 77389-932
1-(3,6-Dichloro-2-fluorophenyl)-2,2-difluoroethanone ≥95%
1-(3,6-Dichloro-2-fluorophenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₈H₃Cl₂F₃O
MW: 243.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762351
CAS Number: 2807466-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR