1-(2,3-Dichloro-6-fluorophenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

65986-500MG 65986-1G 65986-5G 65986-25G
77332-698EA 258.53 USD
77332-698 77356-954 77368-744 77389-930
1-(2,3-Dichloro-6-fluorophenyl)-2,2-difluoroethanone ≥97%
1-(2,3-Dichloro-6-fluorophenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₈H₃Cl₂F₃O
MW: 243.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762350
CAS Number: 2807465-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR