1-(3-Chloro-2,6-difluorophenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

65984-500MG 65984-1G 65984-5G 65984-25G
77332-696EA 258.53 USD
77332-696 77356-952 77368-742 77389-928
1-(3-Chloro-2,6-difluorophenyl)-2,2-difluoroethanone ≥97%
1-(3-Chloro-2,6-difluorophenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₈H₃ClF₄O
Storage Temperature: Ambient
MDL Number: MFCD34762349
CAS Number: 2229138-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR