1-(2-Chloro-6-fluorophenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

65983-500MG 65983-1G 65983-5G
77335-448EA 353.29 USD
77335-448 77356-950 77380-898
1-(2-Chloro-6-fluorophenyl)-2,2-difluoroethanone ≥97%
1-(2-Chloro-6-fluorophenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O
MW: 208.565 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762348
CAS Number: 1865063-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR